siRNA Details
Gene Symbol: Olr1091 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000593.1 | NP_001000593.1 | 1 | 754 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
