siRNA Details
Gene Symbol: Tbc1d15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241353.2 | XP_006241415.1 | 14 | 1614 | ||
XM_345825.7 | XP_345826.3 | 13 | 1563 | |||
GenBank mRNA | BC085348.1 | - | 1 | 28 | ||
BC099146.1 | - | 1 | 90 |

More Information
Other Species Targeted

Gene Ontology Categories:
