siRNA Details
Gene Symbol: Tcf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130574.1 | NP_001124046.1 | 2 | 1061 | ||
XM_006242110.2 | XP_006242172.1 | 1 | 1961 | |||
XM_006242112.2 | XP_006242174.1 | 1 | 1961 | |||
XM_008765772.1 | XP_008763994.1 | 1 | 1960 |

More Information
Other Species Targeted

Gene Ontology Categories:
