siRNA Details
Gene Symbol: Pphln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108992.1 | NP_001102462.1 | 3 | 482 | ||
XM_006242230.1 | XP_006242292.1 | 3 | 479 | |||
XM_006242231.1 | XP_006242293.1 | 3 | 479 | |||
XM_006242236.2 | XP_006242298.1 | 3 | 246 | |||
GenBank mRNA | AY724510.1 | - | 2 | 215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
