siRNA Details
Gene Symbol: Nfe2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012224.1 | NP_001012224.1 | 3 | 755 | ||
XM_006242431.2 | XP_006242493.1 | 4 | 967 | |||
XM_006242432.2 | XP_006242494.1 | 3 | 786 | |||
XM_006242433.2 | XP_006242495.1 | 3 | 1412 | |||
GenBank mRNA | BC078925.1 | - | 3 | 755 | 7106450 | |
FQ233359.1 | - | 3 | 589 | |||
FQ234994.1 | - | 3 | 600 |
