siRNA Details
Gene Symbol: Zfp426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079943.1 | NP_001073412.1 | 8 | 1069 | ||
XM_006242666.1 | XP_006242728.1 | 7 | 1076 | |||
XM_006242667.2 | XP_006242729.1 | 7 | 3164 | |||
XM_006242668.2 | XP_006242730.1 | 8 | 2470 | |||
XM_006242670.1 | XP_006242732.1 | 8 | 1159 | |||
XM_006242671.2 | XP_006242733.1 | 6 | 945 | |||
XM_006242672.2 | XP_006242734.1 | 7 | 2672 | |||
XM_006242673.2 | XP_006242735.1 | 6 | 2525 | |||
XM_008765981.1 | XP_008764203.1 | 7 | 2675 | |||
XM_008765982.1 | XP_008764204.1 | 7 | 1012 | |||
XM_008765983.1 | XP_008764205.1 | 7 | 2320 | |||
XM_008765984.1 | XP_008764206.1 | 7 | 2276 | |||
GenBank mRNA | BC129125.1 | - | 8 | 1069 | 7133611 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
