siRNA Details
Gene Symbol: LOC102550797 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766081.1 | XP_008764303.1 | 692 | |||
XM_008766082.1 | XP_008764304.1 | 692 | ||||
XM_008766083.1 | XP_008764305.1 | 542 |
Gene Symbol: RGD1563738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766092.1 | XP_008764314.1 | 4 | 692 | ||
XM_008766093.1 | XP_008764315.1 | 4 | 692 | |||
XM_008766095.1 | XP_008764317.1 | 3 | 542 |
