siRNA Details
Gene Symbol: Alg9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109000.1 | NP_001102470.1 | 3 | 510 | ||
XM_008766322.1 | XP_008764544.1 | 3 | 1088 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
