siRNA Details
Gene Symbol: Neil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025754.1 | NP_001020925.1 | 9 | 1061 | ||
XM_006243134.2 | XP_006243196.1 | 9 | 1875 | |||
XM_006243135.2 | XP_006243197.1 | 9 | 1981 | |||
XM_006243136.2 | XP_006243198.1 | 10 | 1057 | |||
GenBank mRNA | BC099125.1 | - | 9 | 1061 | 7445605 |
