siRNA Details
Gene Symbol: Gk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766574.1 | XP_008764796.1 | 11 | 1051 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766574.1 | XP_008764796.1 | 11 | 1051 |