siRNA Details
Gene Symbol: Cpne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109003.1 | NP_001102473.1 | 7, 8 | 1270 | ||
XM_008766557.1 | XP_008764779.1 | 4, 5 | 885 | |||
GenBank mRNA | AM747281.1 | - | 3, 4 | 433 |

More Information
Other Species Targeted

