siRNA Details
Gene Symbol: Oard1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244466.2 | XP_006244528.1 | 5 | 946 | ||
XM_006244467.2 | XP_006244529.1 | 5 | 943 | |||
XM_006244470.2 | XP_006244532.1 | 5 | 538 | |||
XM_006244471.2 | XP_006244533.1 | 5 | 771 | |||
XM_006244472.2 | XP_006244534.1 | 5 | 542 | |||
XM_008766872.1 | XP_008765094.1 | 5 | 954 | |||
XM_008766873.1 | XP_008765095.1 | 5 | 952 | |||
XM_008766874.1 | XP_008765096.1 | 6 | 818 | |||
XM_008766875.1 | XP_008765097.1 | 5 | 676 |

More Information
Other Species Targeted

Gene Ontology Categories:
