siRNA Details
Gene Symbol: Oard1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244466.2 | XP_006244528.1 | 4, 5 | 870 | ||
XM_006244467.2 | XP_006244529.1 | 4, 5 | 867 | |||
XM_006244470.2 | XP_006244532.1 | 4, 5 | 462 | |||
XM_006244471.2 | XP_006244533.1 | 4, 5 | 695 | |||
XM_006244472.2 | XP_006244534.1 | 4, 5 | 466 | |||
XM_008766872.1 | XP_008765094.1 | 4, 5 | 878 | |||
XM_008766873.1 | XP_008765095.1 | 4, 5 | 876 | |||
XM_008766874.1 | XP_008765096.1 | 5, 6 | 742 | |||
XM_008766875.1 | XP_008765097.1 | 4, 5 | 600 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_207219, AK148136, BC022574, AK153512, BC157987.
Gene Ontology Categories:
