siRNA Details
Gene Symbol: Runx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278483.1 | NP_001265412.1 | 9 | 1563 | ||
XM_006244549.2 | XP_006244611.1 | 8 | 1764 | |||
XM_006244550.2 | XP_006244612.1 | 9 | 1775 | |||
XM_006244551.2 | XP_006244613.1 | 9 | 1742 | |||
XM_006244554.2 | XP_006244616.1 | 6 | 1170 |

More Information
Other Species Targeted

Gene Ontology Categories:
