siRNA Details
Gene Symbol: Rab23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109005.1 | NP_001102475.1 | 5 | 692 | ||
XM_008766955.1 | XP_008765177.1 | 5 | 1353 | |||
XM_008766956.1 | XP_008765178.1 | 5 | 1358 | |||
XM_008766957.1 | XP_008765179.1 | 5 | 678 | |||
XM_008766958.1 | XP_008765180.1 | 5 | 1020 | |||
XM_008766959.1 | XP_008765181.1 | 5 | 1351 | |||
XM_008766960.1 | XP_008765182.1 | 5 | 1346 | |||
XM_008766961.1 | XP_008765183.1 | 3 | 2069 |
