siRNA Details
Gene Symbol: Mgat4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012225.2 | NP_001012225.1 | 11, 12 | 1404 | ||
NM_001160155.1 | NP_001153627.1 | 11, 12 | 1431 | |||
XM_006244840.2 | XP_006244902.1 | 11, 12 | 1456 | |||
XM_008767040.1 | XP_008765262.1 | 10, 11 | 1119 | |||
GenBank mRNA | BC088215.1 | - | 10, 11 | 1362 | 7383944 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
