siRNA Details
Gene Symbol: Stat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012226.1 | NP_001012226.1 | 6 | 989 | ||
GenBank mRNA | BC081938.1 | - | 6 | 989 | 7115387 |

More Information
Other Species Targeted

Gene Ontology Categories:
