siRNA Details
Gene Symbol: March4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001074008.4 | XP_001074008.1 | 4 | 976 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001045533, BC098191, BC056494, AK122502.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
