siRNA Details
Gene Symbol: Kcne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212526.1 | NP_997691.1 | 2 | 181 | ||
XM_008767283.1 | XP_008765505.1 | 2 | 446 | |||
GenBank mRNA | AF512994.1 | - | 2 | 181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
