siRNA Details
Gene Symbol: Slco6d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014270.1 | NP_001014292.1 | 1845 | |||
XM_006245580.2 | XP_006245642.1 | 2083 | ||||
XM_006245583.2 | XP_006245645.1 | 1822 | ||||
GenBank mRNA | BC083829.1 | - | 1845 | 7112354 |
