siRNA Details
Gene Symbol: Rp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109012.1 | NP_001102482.1 | 2 | 911 | ||
XM_008773035.1 | XP_008771257.1 | 2 | 911 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109012.1 | NP_001102482.1 | 2 | 911 | ||
XM_008773035.1 | XP_008771257.1 | 2 | 911 |