siRNA Details
Gene Symbol: Tmem164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109014.1 | NP_001102484.1 | 5 | 861 | ||
XM_008773426.1 | XP_008771648.1 | 5 | 890 | |||
XM_008773427.1 | XP_008771649.1 | 4 | 749 | |||
XM_008773428.1 | XP_008771650.1 | 4 | 287 |

More Information
Other Species Targeted

