siRNA Details
Gene Symbol: Ube4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271198.1 | NP_001258127.1 | 25 | 4054 | ||
XM_006239387.2 | XP_006239449.1 | 27 | 4588 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
