siRNA Details
Gene Symbol: Zic3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109019.2 | NP_001102489.1 | 1 | 371 | ||
XM_006257586.2 | XP_006257648.1 | 2 | 764 | |||
XM_008773570.1 | XP_008771792.1 | 2 | 784 | |||
XM_008773571.1 | XP_008771793.1 | 2 | 784 |

More Information
Other Species Targeted

Gene Ontology Categories:
