siRNA Details
Gene Symbol: RGD1560171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752150.3 | XP_003752198.1 | 1 | 383 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1560171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752150.3 | XP_003752198.1 | 1 | 383 |