siRNA Details
Gene Symbol: LOC367975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079896.1 | NP_001073365.1 | 5 | 772 | ||
XM_006257480.2 | XP_006257542.1 | 5 | 791 | |||
GenBank mRNA | BC127533.1 | - | 5 | 772 | 7132670 |
Gene Symbol: RGD1563150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257484.2 | XP_006257546.1 | 3 | 403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
