siRNA Details
Gene Symbol: LOC367975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079896.1 | NP_001073365.1 | 5 | 779 | ||
XM_006257480.2 | XP_006257542.1 | 5 | 798 | |||
GenBank mRNA | BC127533.1 | - | 5 | 779 | 7132670 |
Gene Symbol: RGD1563150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257484.2 | XP_006257546.1 | 3 | 410 |
