siRNA Details
Gene Symbol: Pnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109023.1 | NP_001102493.1 | 5, 6 | 443 | ||
GenBank mRNA | FQ234501.1 | - | 5, 6 | 452 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008891, BC132571, AK030978, Y08701, AK050669, and others.
Gene Ontology Categories:
