siRNA Details
Gene Symbol: Pnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109023.1 | NP_001102493.1 | 2, 3 | 206 | ||
GenBank mRNA | FQ234501.1 | - | 2, 3 | 215 |

More Information
Other Species Targeted

Gene Ontology Categories:
