siRNA Details
Gene Symbol: Hpse2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135762.1 | NP_001129234.1 | 13 | 1247 | ||
XM_008760445.1 | XP_008758667.1 | 9 | 3431 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081257, BC158133, BC158131, BC158136.
Gene Ontology Categories:
