siRNA Details
Gene Symbol: Rorc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232926.2 | XP_006232988.2 | 10 | 1377 | ||
XM_008761339.1 | XP_008759561.1 | 9 | 3701 | |||
GenBank mRNA | BC169105.1 | - | 275 | 7383660 |
