siRNA Details
Gene Symbol: Ift81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199120.1 | NP_954551.1 | 17 | 1871 | ||
GenBank mRNA | AY345164.1 | - | 17 | 1871 | ||
FQ225832.1 | - | 17 | 1904 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
