siRNA Details
Gene Symbol: Ift81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199120.1 | NP_954551.1 | 16, 17 | 1813 | ||
GenBank mRNA | AY345164.1 | - | 16, 17 | 1813 |

More Information
Other Species Targeted

Gene Ontology Categories:
