siRNA Details
Gene Symbol: Tmem159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194354.2 | NP_919335.2 | 3, 4 | 270 | ||
XM_006230138.2 | XP_006230200.1 | 2, 3 | 363 | |||
XM_006230139.2 | XP_006230201.1 | 3, 4 | 408 | |||
XM_008759784.1 | XP_008758006.1 | 2, 3 | 1201 | |||
XM_008759785.1 | XP_008758007.1 | 2, 3 | 1196 | |||
GenBank mRNA | AY368496.1 | - | 1, 2 | 104 | ||
BC076384.1 | - | 3, 4 | 270 | 7107584 |

More Information
Other Species Targeted

