siRNA Details
Gene Symbol: Trmt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198051.1 | NP_932168.1 | 9 | 863 | ||
XM_006227748.2 | XP_006227810.1 | 9 | 877 | |||
XM_006227749.2 | XP_006227811.1 | 9 | 1394 | |||
XM_006227750.2 | XP_006227812.1 | 9 | 876 | |||
XM_006227751.2 | XP_006227813.1 | 9 | 875 | |||
XM_006227752.2 | XP_006227814.1 | 9 | 875 | |||
XM_008758673.1 | XP_008756895.1 | 9 | 1537 | |||
GenBank mRNA | AF532978.1 | - | 9 | 863 |

More Information
Other Species Targeted

Gene Ontology Categories:
