siRNA Details
Gene Symbol: aicda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100779.1 | NP_001094249.1 | 3, 4 | 518 | ||
GenBank mRNA | AJ606071.2 | - | 2, 3 | 339 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
