siRNA Details
Gene Symbol: Mrgpre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002288.1 | NP_001002288.1 | 1 | 265 | ||
XM_006230870.2 | XP_006230932.1 | 4 | 831 | |||
XM_006230872.2 | XP_006230934.1 | 2 | 629 | |||
XM_008760163.1 | XP_008758385.1 | 3 | 727 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
