siRNA Details
Gene Symbol: Olr733 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000618.1 | NP_001000618.1 | 1 | 629 |
Gene Symbol: Olr736 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001384.1 | NP_001001384.1 | 1 | 629 |
Gene Symbol: Olr745 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000578.1 | NP_001000578.1 | 1 | 629 |

More Information
Other Species Targeted

Gene Ontology Categories:
