siRNA Details
Gene Symbol: Olr727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000619.1 | NP_001000619.1 | 1 | 238 |
Gene Symbol: Olr726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000620.2 | NP_001000620.2 | 2 | 178 |
Gene Symbol: Olr728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000363.1 | NP_001000363.1 | 1 | 235 |

More Information
Other Species Targeted

Gene Ontology Categories:
