siRNA Details
Gene Symbol: Olr582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000662.1 | NP_001000662.1 | 1 | 303 | ||
XM_006234491.1 | XP_006234553.1 | 1 | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
