siRNA Details
Gene Symbol: Olr496 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000681.1 | NP_001000681.1 | 1 | 423 |
Gene Symbol: LOC100912505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749500.1 | XP_003749548.1 | 1 | 423 |
