siRNA Details
Gene Symbol: Olr1496 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000716.1 | NP_001000716.1 | 1 | 664 | ||
GenBank mRNA | M64385.1 | - | 1 | 664 |
Gene Symbol: LOC100909578 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750853.1 | XP_003750901.1 | 1 | 664 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
