siRNA Details
Gene Symbol: Olr1470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000723.1 | NP_001000723.1 | 1 | 578 | ||
GenBank mRNA | M64388.1 | - | 1 | 578 |
Gene Symbol: LOC100909493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750851.3 | XP_003750899.1 | 1 | 578 |

More Information
Other Species Targeted

Gene Ontology Categories:
