siRNA Details
Gene Symbol: Olr1469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212527.1 | NP_997692.1 | 1 | 592 | ||
GenBank mRNA | M64375.1 | - | 1 | 16 | ||
Y07557.1 | - | 1 | 592 |
Gene Symbol: LOC100909534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750852.1 | XP_003750900.1 | 1 | 592 |
Gene Symbol: Olr1482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000026.1 | NP_001000026.1 | 1 | 592 | ||
XM_008768133.1 | XP_008766355.1 | 1 | 592 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
