siRNA Details
Gene Symbol: Olr241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000734.1 | NP_001000734.1 | 1 | 237 |
Gene Symbol: Olr240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229963.2 | XP_006230025.1 | 1 | 130 |
