siRNA Details
Gene Symbol: Olr57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000749.1 | NP_001000749.1 | 1 | 411 |

More Information
Other Species Targeted

Gene Ontology Categories:

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000749.1 | NP_001000749.1 | 1 | 411 |