siRNA Details
Gene Symbol: Olr331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000760.1 | NP_001000760.1 | 1 | 898 |
Gene Symbol: Olr387 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000109.1 | NP_001000109.1 | 1 | 889 |
