siRNA Details
Gene Symbol: Olr1274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000801.1 | NP_001000801.1 | 1 | 517 |
Gene Symbol: LOC100909993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749181.1 | XP_003749229.1 | 517 |
Gene Symbol: LOC100910051 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749261.1 | XP_003749309.1 | 1 | 517 |
Gene Symbol: LOC100910199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749264.1 | XP_003749312.1 | 517 |
Gene Symbol: LOC100910402 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749187.1 | XP_003749235.1 | 1 | 517 |
