siRNA Details
Gene Symbol: Olr1262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000803.1 | NP_001000803.1 | 1 | 393 |
Gene Symbol: LOC102552923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235498.1 | XP_006235560.1 | 1 | 393 |
Gene Symbol: Olr1261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000804.1 | NP_001000804.1 | 1 | 393 |
