siRNA Details
Gene Symbol: Olr1239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000811.1 | NP_001000811.1 | 1 | 490 |
Gene Symbol: LOC103692040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762674.1 | XP_008760896.1 | 2 | 508 |
